missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Toxoplasma Gondii T. gondii MIC-3 Protein
A cDNA sequence encoding the T. gondii MIC-3 was constructed and used to recombinantly synthesize the protein.
£173.00 - £1100.00
Specifications
Name | Toxoplasma Gondii T. gondii MIC-3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Toxoplasma Gondii |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944309
|
enQuireBio™
QP13663-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964309
|
enQuireBio™
QP13663-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954309
|
enQuireBio™
QP13663-1MG |
1 mg |
£1100.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Toxoplasma Gondii T. gondii MIC-3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Toxoplasma Gondii | |
Untagged | |
Toxoplasma protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl pH 8.0, 1.5M urea and 50% glycerol. |