missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Sheep IFN-tau-1 Protein

Product Code. p-200083644 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
This item is not returnable. View return policy

Product Code. 18601397

Brand: Novus Biologicals™ NBP2614401000ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

IFN-tau-1 protein is fully biologically active when compared to IFN-alpha. The specific activity determined by a viral resistance assay is no less than 1.0 x 10^7 IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 100144750
Molecular Weight (g/mol) MolecularWeight-theroretical: 19.9 kDa
Quantity 1000 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 0.1 EU/μg of IFN-tau-1 AS determined by LAL method.
Gene Alias Antiluteolysin, interferon tau-1, TP-1, Trophoblast antiluteolytic protein, Trophoblast protein 1, Trophoblastin
Gene Symbol TP-1
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Purity or Quality Grade >97 % pure by SDS-PAGE and HPLC
Protein IFN-tau-1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.