missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant SARS SARS Core Protein
A cDNA sequence encoding the SARS Core was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10499-1mg
Additional Details : Weight : 0.01000kg
Specifications
SARS SARS Core Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, 60mM NaCl and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
SARS | |
Untagged | |
SARS Core protein is >95% pure as determined by 10% PAGE (coomassie staining). |