missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant SARS-CoV Spike (1-53, 90-115, 171-205 aa) His (C-Term) Protein

Product Code. 18328544 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328544 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328544 Supplier Novus Biologicals™ Supplier No. NBP307937

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Formulation PBS
Quantity 0.025 mg
Storage Requirements Store at -20 to -70 C. Avoid freeze-thaw cycles.
Purity or Quality Grade >90%, by SDS-PAGE
Protein Spike
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.