missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant SARS-CoV-2 Spike RBD (E484K) His (C-Term) Protein

Product Code. 18375225 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375225 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375225 Supplier Novus Biologicals™ Supplier No. NBP307926

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Formulation ddH2O
Molecular Weight (g/mol) M.W. Theoretical: 35 kDa
Quantity 0.05 mg
Storage Requirements Store at 4 C short term. Store (at -20 C long term. Avoid freeze-thaw cycles.)
Gene Alias 501 Y.V2/V3 variant
Purity or Quality Grade >90%, by SDS-PAGE
Protein Spike RBD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.