missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant SARS-CoV-2 Nucleocapsid His (C-Term) Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
Specifications
Specifications
| Formulation | PBS and 10 mM K2CO3. |
| Gene ID (Entrez) | 43740575 |
| Quantity | 0.05 mg |
| Storage Requirements | Store at -20°C. Avoid freeze/thaw cycles. |
| Gene Alias | 2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein |
| Conjugate | Unconjugated |
| Species | SARS-CoV-2 |
| Recombinant | Recombinant |
| Form | Purified |
| Research Category | Virology Bacteria and Parasites |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction