missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant SARS-CoV-2 K986P V987P Spike S2 (GCN4-IZ) His CF
Description
Bioassay data are not available.
Specifications
Specifications
| Accession Number | YP_009724390.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 43740568 |
| Molecular Weight (g/mol) | M.W. (Observed): 75-95 kDa, under reducing conditions, M.W. (Theoretical): 63 kDa |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike S2 Subunit protein SARS-CoV-2 Spike S2 Subunit (Ser686-Lys1211)(Lys986Pro, Val987Pro) (N-terminus) Accession # YP_009724390.1 GCN4-IZ HHHHHH (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | COVID-19, S2 protein, SARS-CoV-2, SARS-CoV-2 Spike, Spike glycoprotein Subunit 2, spike protein, Spike protein S2, structural protein, surface glycoprotein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?