missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant SARS-CoV-2 C.37 S (GCN4-IZ) Avi His Protein

Product Code. 18778156
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18778156 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18778156 Supplier R&D Systems™ Supplier No. AVI10887050
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Avi-tag His-tag Lambda Variant

Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 Fc Chimera (Catalog # 10544-ZN ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number YP_009724390.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 43740568
Molecular Weight (g/mol) M.W.-observed: 145 - 170 kDa, under reducing conditions., M.W.-theroretical: 137 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike protein SARS-CoV-2 C0.37 Spike(Val16-Lys1211)(Gly75Val, Thr76Ile, RSYLTPG246-252 del, Asp253Asn, Leu452Gln, Phe490Ser, Asp614Gly, Thr859Asn) (Arg682Ser, Arg685Ser, Lys986Pro, Val987Pro)(N-terminus) Accession # YP_009724390.1 GCN4-IZ Avi-tag 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S Protein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein
Conjugate Biotin
Reconstitution Reconstitute at 500 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein SARS-CoV-2 Spike
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.