missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant SARS-CoV-2 AV.1 Spike (GCN4-IZ) His Protein

Product Code. 18726826
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18726826

Brand: R&D Systems™ 10896CV100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 His-tag (Catalog # 933-ZN ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number YP_009724390.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 43740568
Molecular Weight (g/mol) M.W.-observed: 150 - 172 kDa, under reducing conditions., M.W.-theroretical: 138 kDa
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike protein SARS-CoV-2 AV0.1 Spike (Val16-Lys1211)(Asp80Gly, Thr95Ile, Gly142Asp, Tyr144 del, Asn439Lys, Glu484Lys, Asp614Gly, Pro681His, Ile1130Val, Asp1139His) (Arg682Ser, Arg685Ser, Lys986Pro, Val987Pro) (N-terminus) Accession # YP_009724390.1 GCN4-IZ 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S Protein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein SARS-CoV-2 Spike
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.