missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Salmonella Typhi S. Typhi OMP 52kDa Protein
A cDNA sequence encoding the S. Typhi OMP 52kDa was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13385-1mg
Additional Details : Weight : 0.01000kg
Specifications
Salmonella Typhi S. Typhi OMP 52kDa Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized from 1 mg/ml in 20mM sodium carbonate pH 10. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Salmonella Typhi | |
His | |
Greater than 95.0% as determined by SDS-PAGE. |