missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Salmonella Typhi S. Typhi H Protein
A cDNA sequence encoding the S. Typhi H was constructed and used to recombinantly synthesize the protein.
£173.00 - £1320.00
Specifications
Name | Salmonella Typhi S. Typhi H Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Salmonella Typhi |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15953389
|
enQuireBio™
QP13382-100ug |
100 μg |
£173.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15973389
|
enQuireBio™
QP13382-500ug |
500 μg |
£660.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15963389
|
enQuireBio™
QP13382-1mg |
1 mg |
£1320.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Salmonella Typhi S. Typhi H Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Salmonella Typhi | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile Filtered solution containing PBS and 25mM arginine. |