missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Salamander Leptin Salamander Protein

Product Code. 15915088
Click to view available options
Quantity:
1 mg
100 μg
200 μg
Unit Size:
100µg
1mg
200µg
This item is not returnable. View return policy

Product Code. 15915088

Brand: enQuireBio™ QP10760100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Leptin Salamander was constructed and used to recombinantly synthesize the protein.

Specifications

Name Salamander Leptin Salamander Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity By inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Its activity in that test is similar to that of mouse leptin (see Salomon et al (2006) Protein Expression and Purifcation 47, 128–136).
Product Type Recombinant Protein
Cross Reactivity Salamander
Species E. coli
Protein Tag Untagged
Sequence The sequence of the ten N-terminal amino acids was determined and was found to be Ala-Ile-Met-Val-Asp-Gln-Leu-Arg-Met-Asp
Buffer The Salamander Leptin was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
Purity or Quality Grade Greater than 99.0% as determined by: (a) Analysis by Gel-Filtration. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.