missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant S. plicatus Endo H Protein
Description
Measured by its ability to deglycosylate ribonuclease B under native conditions. The DC50 is <5 ng. The DC50 is defined as the amount of enzyme required to remove 50% of glycan on 1 μg of RNase B in 30 minutes at 37 °C. . Use of Recombinant S. plicatus Endo-beta-N-acetylglucosaminidase H/Endo H in the deglycoslyation of other substrates may require alternative conditions for optimal performance.
Specifications
Specifications
| Accession Number | P04067 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and EDTA. |
| Molecular Weight (g/mol) | M.W. (Observed): 29 kDa, reducing conditions; M.W. (theoretical): 30 kDa |
| Name | Endo-beta-N-acetylglucosaminidase H/Endo H |
| Quantity | 50 μg |
| Source | E. coli-derived s. plicatus Endo-beta-N-acetylglucosaminidase H/Endo H protein Val47-Pro313 with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.01 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?