missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae Ubiquitin-like protein SMT3 Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Ubiquitin-like protein SMT3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7546-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049310

  • £1930.00 / 500µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_010798.1
S. cerevisiae Ubiquitin-like protein SMT3 Protein
Research Use Only
SMT3
Recombinant Protein
Yeast
SDSEVNQEAK PEVKPEVKPE THINLKVSDG SSEIFFKIKK TTPLRRLMEA FAKRQGKEMD SLRFLYDGIR IQADQTPEDL DMEDNDIIEA HREQIGG
Greater than 90% as determined by SDS-PAGE.
852122
500 μg
Not determined.
Full Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers