missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae Ubiquitin-like protein SMT3 Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Ubiquitin-like protein SMT3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7546-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029310

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_010798.1
S. cerevisiae Ubiquitin-like protein SMT3 Protein
Research Use Only
SMT3
Recombinant Protein
Yeast
SDSEVNQEAK PEVKPEVKPE THINLKVSDG SSEIFFKIKK TTPLRRLMEA FAKRQGKEMD SLRFLYDGIR IQADQTPEDL DMEDNDIIEA HREQIGG
Greater than 90% as determined by SDS-PAGE.
852122
1 mg
Not determined.
Full Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers