missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Pyruvate carboxylase 1 Protein
Click to view available options
Quantity:
1 mg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
1mg
200µg
500µg
50µg
Specifications
Specifications
| Accession Number | NP_011453.1 |
| Gene ID (Entrez) | 852818 |
| Name | S. cerevisiae Pyruvate carboxylase 1 Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not determined. |
| Gene Symbol | PYC1 |
| Protein Length | Partial (see sequence information for more details). |
| Product Type | Recombinant Protein |
| Cross Reactivity | S. cerevisiae |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction