missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae 26S proteasome regulatory subunit RPN13 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) 26S proteasome regulatory subunit RPN13, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7460-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_013525.3 | |
S. cerevisiae 26S proteasome regulatory subunit RPN13 Protein | |
Research Use Only | |
RPN13 | |
Recombinant Protein | |
E. coli | |
SMSSTVIKFR AGVCEYNEDS RLCTPIPVQG EIEIKPNEEE ELGFWDFEWR PTEKPVGREL DPISLILIPG ETMWVPIKSS KSGRIFALVF SSNERYFFWL QEKNSGNLPL NELSAKDKEI YNKMIGVLNN SSESDEEESN DEKQKAQDVD VSMQD | |
Greater than 90% as determined by SDS-PAGE. |
851140 | |
500 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |