missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae 26S proteasome regulatory subunit RPN13 Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) 26S proteasome regulatory subunit RPN13, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7460-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048910

  • £715.00 / 100µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_013525.3
S. cerevisiae 26S proteasome regulatory subunit RPN13 Protein
Research Use Only
RPN13
Recombinant Protein
E. coli
SMSSTVIKFR AGVCEYNEDS RLCTPIPVQG EIEIKPNEEE ELGFWDFEWR PTEKPVGREL DPISLILIPG ETMWVPIKSS KSGRIFALVF SSNERYFFWL QEKNSGNLPL NELSAKDKEI YNKMIGVLNN SSESDEEESN DEKQKAQDVD VSMQD
Greater than 90% as determined by SDS-PAGE.
851140
100 μg
Not determined.
Full Length
S. cerevisiae
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers