missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Prostatic Acid Phosphatase / ACPP Protein

A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) Prostatic Acid Phosphatase / ACPP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6412-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003130

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_000426914.1
S. aureus Prostatic Acid Phosphatase / ACPP Protein
Research Use Only
acpP
Recombinant Protein
E. coli
MENFDKVKDI IVDRLGVDAD KVTEDASFKD DLGADSLDIA ELVMELEDEF GTEIPDEEAE KINTVGDAVK FINSLEK
Greater than 90% as determined by SDS-PAGE.
31213993
500 μg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers