missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Gamma-hemolysin component C Protein

Product Code. p-7154413
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16070501

Brand: enQuireBio™ QP7736ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Staphylococcus aureus (strain MRSA252) Gamma-hemolysin component C, was expressed in the hosts and tags indicated.

Specifications

Accession Number WP_000916697.1
Name S. aureus Gamma-hemolysin component C Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol hlgC
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity S. aureus
Species E. coli
Protein Tag His-SUMO
Sequence ANDTEDIGKG NDVEIIKRTE DKTSNKWGVT QNIQFDFVKD KKYNKDALIL KMQGFISSRT TYYNYKNTNH IKSMRWPFQY NIGLKTNDKY VSLINYLPKN KIESTNVSQT LGYNIGGNFQ SAPSLGGNGS FNYSKSISYT QQNYVSEVEQ QNSKSVLWGV KANSFATESG QKSAFDSDLF VGYKPHSKDP RDYFVPDSEL PPLVQSGFNP SFIATVSHEK GSSDTSEFEI TYGRNMDVTH AIKRSTHYGN SYLDGHRVHN AFKNRNYTVK YEVNWKTHEI KVKGQN
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.