missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rotavirus A Outer capsid protein VP4 Protein
Click to view available options
Quantity:
1 mg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
1mg
200µg
500µg
50µg
Specifications
Specifications
| Name | Rotavirus A Outer capsid protein VP4 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not determined. |
| Protein Length | Full Length |
| Product Type | Recombinant Protein |
| Cross Reactivity | Rotavirus |
| Species | E. coli |
| Protein Tag | His-SUMO |
| Sequence | AQVSEDIIIS KTSLWKEMQY NRDIIIRFKF NNSIIKLGGL GYKWSEISFK AANYQYNYLR DGEQVTAHTT CSVNGVNNFS YNGGLLPTHF SISRYEVIKE NSYVYVDYWD DSQAFRNMVY VRSLAANLNS VKCSGGNYNF QMPVGAWPVM SGGAVSLHFA GVTLSTQFTD FVSLNSLRFR FSLTVEEPPF SILRTRVSGL YGLPASNPNS GHEYYEIAGR FSLISLVPSN DDY |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction