missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rhesus Macaque IL 13 Protein

A cDNA sequence encoding the IL 13 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10691-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944668

  • £99.15 / 2µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

IL 13 Protein
Research Use Only
Recombinant Protein
E. coli
SPSPVPRSTA LKELIEELVN ITQNQKAPLC NGSMVWSINL TAGVYCAALE SLINVSGCSA IEKTQRMLNG FCPHKVSAGQ FSSLRVRDTK IEVAQFVKDL LVHLKKLFRE GRFN
Greater than 97.0% as determined by SDS-PAGE and HPLC analyses.
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
Rhesus Macaque
Untagged
Lyophilized from a 0.2?m filtered concentrated solution in PBS, pH 7.4 containing 5% trehalose.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers