missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rhesus Macaque IL-1 beta/IL-1F2 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
10µg
Description
The Recombinant Rhesus Macaque IL-1 beta/IL-1F2 Protein is derived from E. coli. The Recombinant Rhesus Macaque IL-1 beta/IL-1F2 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Molecular Weight (g/mol) | 17.3 kDa |
| Name | IL-1 beta/IL-1F2 |
| Quantity | 10 μg |
| Source | E. coli-derived rhesus macaque IL-1 beta/IL-1F2 protein Ala117-Ser269 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction