missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rhesus Macaque IL-1 beta/IL-1F2 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
Description
IL1 beta Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine D10.G4.1 cells is less than 10 pg/mL, corresponding to a specific activity of > 1.0 x 10^8 IU/mg.
Specifications
Specifications
| Accession Number | P48090, P48090, P48090 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 3553 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 17.3 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of IL-1 beta/IL-1F2 as determined by LAL method. |
| Gene Alias | catabolin, IL-1, IL-1 beta, IL-1 B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta |
| Gene Symbol | IL1B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction