missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque Flt3-Ligand Protein
A cDNA sequence encoding the Flt3-Ligand was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10617-2ug
Additional Details : Weight : 0.01000kg
Specifications
Flt3-Ligand Protein | |
Research Use Only | |
The ED50 as calculated by the dose-dependant stimulation of the proliferation of human AML5 cells is less than 1.0ng/ml, corresponding to a specific activity of 1,000,000IU/mg. | |
Rhesus Macaque | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
TQDCSFQHSP ISSDFAVKIR ELSDYLLQDY PVTVPSNLQD EELCGALWRL VLAQRWMERL KTVAGSKMQG LLERVNTEIH FVTKCAFQHP PSCLRFVQTN ISRLLQETSE QLVALKPWIT RQNFSRCLEL QCQPDSSTLP PPRSPGALEA TALTAPQRP | |
Greater than 97.0% as determined by SDS-PAGE. |