missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant RFP His (N-Term) Protein

Product Code. 18066098 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18066098

Brand: Novus Biologicals™ NBP199583100UG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

Recombinant protein for Human RFP The Recombinant Non-species specific RFP Protein is derived from E. coli. The Recombinant Non-species specific RFP Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Lyoph
For Use With (Application) SDS-PAGE
Formulation Freeze Dried
Molecular Weight (g/mol) 27.6 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias dsRedGene Symbol, Recombinant RFP Protein, Red Fluorescent Protein
Reconstitution Reconstitute with dH2O to 1 mg/mL
Buffer Freeze Dried
Endotoxin Level <0.1 ng/μg
Purity or Quality Grade >97%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.