missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rat TRANCE/RANK L/TNFSF11 Protein

Product Code. 18764937
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA.
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18764937

Brand: R&D Systems™ 9366TN025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Rat TRANCE/RANK L/TNFSF11 Protein is derived from CHO. The Recombinant Rat TRANCE/RANK L/TNFSF11 Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number XP_008769150
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA.
Gene ID (Entrez) 8600
Molecular Weight (g/mol) 29 kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived rat TRANCE/TNFSF11/RANK L protein Arg72-Asp318,with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CD254, CD254 antigen, ODF, OPGL, OPGLOPTB2
Biological Activity 2.5 to 15ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.