missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SIRPA Protein
A cDNA sequence encoding the SIRPA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13507-2ug
Additional Details : Weight : 0.01000kg
Specifications
SIRPA Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
KELKVTQADK SVSVAAGDSA TLNCTVSSLT PVGPIKWFKG EGQNRSPIYS FIGGEHFPRI TNVSDATKRN NMDFSICISN VTPEDAGTYY CVKFQKGIVE PDTEIKSGGG TTLYVLAKPS SPEVSGPDSR GSPGQTVNFT CKSYGFSPRN ITLKWLKNGK ELSHLETTIS SKSNVSYNIS STVSVKLSPE DIHSRVICEV AHVTLEGRPL NGTANFSNII RVSPTLKITQ QPLTPASQVN LTCQVQKFYP KALQLNWLEN GNLSRTDKPE HFTDNRDGTY NYTSLFLVNS SAHREDVVFT CQVEHDSQPA ITENHTVRAF AHSSSGGSME TIPDNNAYYN WNVEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
His | |
SIRPA protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |