missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat SCF Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£219.22
Specifications
Gene ID (Entrez) | 60427 |
---|---|
Molecular Weight (g/mol) | 18.5 kDa (monomer) |
Name | SCF/c-kit Ligand |
Quantity | 25μg |
Source | E. coli-derived rat SCF/c-kit Ligand protein Gln26-Ala189,with an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18176360
|
R&D Systems™
6998-SC-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£219.22
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18187000
|
R&D Systems™
6998-SC-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£219.22
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Rat SCF Protein is derived from E. coli. The Recombinant Rat SCF Protein has been validated for the following applications: Bioactivity.Specifications
60427 | |
SCF/c-kit Ligand | |
E. coli-derived rat SCF/c-kit Ligand protein Gln26-Ala189,with an N-terminal Met | |
c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor | |
Recombinant |
18.5 kDa (monomer) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |