missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Rat Progranulin/PGRN FLAG Protein

Product Code. 18399145 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399145 50 μg 50µg
18312312 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18399145 Supplier Novus Biologicals™ Supplier No. NBP31814750UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_058809
Concentration LYOPH
For Use With (Application) SDS-PAGE, Western Blot
Formulation Lyophilized from 0.2um-filtered solution in PBS
Gene ID (Entrez) 2896
Molecular Weight (g/mol) M.W. Theoretical: 65 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.1 EU/μg purified protein (LAL test; Lonza).
Gene Alias CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN
Gene Symbol GRN
Reconstitution Centrifuge lyophilized vial before opening and reconstitution. PBS containing at least 0.1% BSA should be used for further dilutions.
Research Category Cancer, Lipid and Metabolism
Purity or Quality Grade >95% pure by SDS-PAGE
Protein Progranulin/PGRN
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.