missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat PKLR / PKRL Protein

A DNA sequence encoding the Rattus norvegicus (Rat) PKLR / PKRL, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6498-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003750

  • £1600.00 / 500µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_036756.3
PKLR / PKRL Protein
Research Use Only
Pklr
Recombinant Protein
Yeast
MSVQENTLPQ QLWPWIFRSQ KDLAKSALSG APGGPAGYLR RASVAQLTQE LGTAFFQQQQ LPAAMADTFL EHLCLLDIDS QPVAARSTSI IATIGPASRS VDRLKEMIKA GMNIARLNFS HGSHEYHAES IANIREATES FATSPLSYRP VAIALDTKGP EIRTGVLQGG PESEVEIVKG SQVLVTVDPK FQTRGDAKTV WVDYHNITRV VAVGGRIYID DGLISLVVQK IGPEGLVTEV EHGGILGSRK GVNLPNTEVD LPGLSEQDLL DLRFGVQHNV DIIFASFVRK ASDVLAVRDA LGPEGQNIKI ISKIENHEGV KKFDEILEVS DGIMVARGDL GIEIPAEKVF LAQKMMIGRC NLAGKPVVCA TQMLESMITK ARPTRAETSD VANAVLDGAD CIMLSGETAK GSFPVEAVMM QHAIAREAEA AVYHRQLFEE LRRAAPLSRD PTEVTAIGAV EASFKCCAAA IIVLTKTGRS AQLLSQYRPR AAVIAVTRSA QAARQVHLSR GVFPLLYREP PEAIWADDVD RRVQFGIESG KLRGFLRVGD LVIVVTGWRP GSGYTNIMRV LSVS
Greater than 90% as determined by SDS-PAGE.
24651
500 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers