missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PDGF BB Protein
A cDNA sequence encoding the PDGF BB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10805-2ug
Additional Details : Weight : 0.01000kg
Specifications
PDGF BB Protein | |
Research Use Only | |
The ED50as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 2.0 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg. | |
Rat | |
Untagged | |
PDGF-BB was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.0. |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
SLGSLAAAEP AVIAECKTRT EVFQISRNLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RASQVQMRPV QVRKIEIVRK KPVFKKATVT LEDHLACKCE TVVTPRPVT | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |