missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PDGF AA Protein
A cDNA sequence encoding the PDGF AA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10803-10ug
Additional Details : Weight : 0.01000kg
Specifications
PDGF AA Protein | |
Research Use Only | |
The ED50, as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 5.0 ng/ml, corresponding to a specific activity of > 2.0 M 105 IU/mg. | |
Rat | |
Untagged | |
PDGF-AA was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.0. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MSIEEAIPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATSNLN PDHREEETDV R | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |