missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat p53 / TP53 Protein

Product Code. 16076731
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16076731

Brand: enQuireBio™ QP8835ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Rattus norvegicus (Rat) p53 / TP53, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_112251.2
Gene ID (Entrez) 24842
Name p53 / TP53 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol TP53
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag His
Sequence MEDSQSDMSI ELPLSQETFS CLWKLLPPDD ILPTTATGSP NSMEDLFLPQ DVAELLEGPE EALQVSAPAA QEPGTEAPAP VAPASATPWP LSSSVPSQKT YQGNYGFHLG FLQSGTAKSV MCTYSISLNK LFCQLAKTCP VQLWVTSTPP PGTRVRAMAI YKKSQHMTEV VRRCPHHERC SDGDGLAPPQ HLIRVEGNPY AEYLDDRQTF RHSVVVPYEP PEVGSDYTTI HYKYMCNSSC MGGMNRRPIL TIITLEDSSG NLLGRDSFEV RVCACPGRDR RTEEENFRKK EEHCPELPPG SAKRALPTST SSSPQQKKKP LDGEYFTLKI RGRERFEMFR ELNEALELKD ARAAEESGDS RAHSSYPKTK KGQSTSRHKK PMIKKVGPDS D
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.