missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat Neuropilin-1 Fc Chimera (NS0-expressed)
Description
The Recombinant Rat Neuropilin-1 Fc Chimera (NS0-expressed) is derived from NS0. The Recombinant Rat Neuropilin-1 Fc Chimera (NS0-expressed) has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Citrate Phosphate and NaCl. |
| Gene ID (Entrez) | 246331 |
| Molecular Weight (g/mol) | 119 kDa (monomer) |
| Name | Neuropilin-1 Fc Chimera |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived rat Neuropilin-1 protein Rat Neuropilin-1 (Phe22-Ala810) Accession &Num; Q9QWJ9 R Rat Neuropilin-1 (Ser829-Asp854) Accession &Num; Q9QWJ9 IEGRDMD Human IgG1 (Pro100-Lys330) 6-His tag N-terminus C-terminus R&D Systems rat Npn-1 may represent a previously unreported splice isoform of rat Npn-1. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?