missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Midkine Protein
A cDNA sequence encoding the Midkine was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10787-5ug
Additional Details : Weight : 0.01000kg
Specifications
Midkine Protein | |
Research Use Only | |
The biological activity determined by a chemotaxis bioassay using human neutrophils is in a concentration range of 10-100 ng/ml. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
VAKKKDKVKK GSECSEWTWG PCTPSSKDCG MGFREGTCGA QTQRIHCKVP CNWKKEFGAD CKYKFESWGA CDGSTGTKAR QGTLKKARYN AQCQETIRVT KPCTSKTKSK AKAKKGKGKD | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |