missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat MDC Protein
A cDNA sequence encoding the MDC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10783-20ug
Additional Details : Weight : 0.01000kg
Specifications
MDC Protein | |
Research Use Only | |
MDC | |
Recombinant Protein | |
E. coli | |
GPYGANVEDS ICCQDYIRHP LPPRFVKEFY WTSKSCRKPG VVLITIKNRD ICADPRMLWV KKILHKLA | |
Greater than 97.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Determined by its ability to chemoattract human activated lymphocytes using a concentration range of 10.0-100.0 ng/ml. | |
Rat | |
Untagged | |
The protein was lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4. |