missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat MAPT / Tau Protein

Product Code. 16082620
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16082620

Brand: enQuireBio™ QP6345ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Rattus norvegicus (Rat) MAPT / Tau, was expressed in the hosts and tags indicated.

Specifications

Name MAPT / Tau Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Mapt
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag His
Sequence AEPRQEFDTM EDQAGDYTML QDQEGDMDHG LKESPPQPPA DDGSEEPGSE TSDAKSTPTA EDVTAPLVEE RAPDKQATAQ SHTEIPEGTT AEEAGIGDTP NMEDQAAGHV TQEPQKVEIF SQSLLVEPGR REGQAPDSGI SDWTHQQVPS MSGAPLPPQG LREATHQPLG TRPEDVERSH PASELLWQES PQKEAWGKDR LGSEEEVDED ITMDESSQES PPSQASLAPG TATPQARSVS ASGVSGETTS IPGFPAEGSI PLPADFFSKV SAETQASPPE GPGTGPSEEG HEAAPEFTFH VEIKASAPKE QDLEGATVVG APAEEQKARG PSVGKGTKEA SLLEPTDKQP AAGLPGRPVS RVPQLKARVA GVSKDRTGND EKKAKTSTPS CAKTPSNRPC LSPTRPTPGS SDPLIKPSSP AVCPEPATSP KYVSSVTPRN GSPGTKQMKL KGADGKTGAK IATPRGAATP GQKGTSNATR IPAKTTPSPK TPPGSGEPPK SGERSGYSSP GSPGTPGSRS RTPSLPTPPT REPKKVAVVR TPPKSPSASK SRLQTAPVPM PDLKNVRSKI GSTENLKHQP GGGKVQIINK KLDLSNVQSK CGSKDNIKHV PGGGSVHIVY KPVDLSKVTS KCGSLGNIHH KPGGGQVEVK SEKLDFKDRV QSKIGSLDNI THVPGGGNKK IETHKLTFRE NAKAKTDHGA EIVYKSPVVS GDTSPRHLSN VSSTGSIDMV DSPQLATLAD EVSASLAKQG L
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.