missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat M-CSF / CSF-1 Protein
Click to view available options
Quantity:
1 μg
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
1µg
500µg
Specifications
Specifications
| Name | M-CSF / CSF-1 Protein |
| Quantity | 1 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | CSF1 |
| Biological Activity | Rat M-CSF / CSF-1 induces NFS-60 cell proliferation (typical ED50 is ? 2 ng/ml). |
| Product Type | Recombinant Protein |
| Cross Reactivity | Rat |
| Species | E. coli |
| Protein Tag | Untagged |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction