missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Leptin tA Protein
A cDNA sequence encoding the Leptin tA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10765-5ug
This item is not returnable.
View return policy
Specifications
Leptin tA Protein | |
Research Use Only | |
Leptin Antagonist Triple Mutant Rat Recombinant half-life in circulation after SC injection was over 20 hours. Leptin Antagonist Triple Mutant Rat Recombinant is capable of inhibiting leptin-induced proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Leptin Antagonist Triple Mutant Rat Recombinant in vitro activity is 5-6 fold lower than the non-pegylated antagonist, though in vivo it has profound weight gain effect (as compared to the non-pegylated antagonist), resulting mainly from increased food intake. | |
Rat | |
Untagged | |
Greater than 99.0% as determined by:(a) Gel filtration analysis. (b) Analysis by SDS-PAGE. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The Rat Leptin triple antagonist was lyophilized from a concentrated (0.65 mg/ml) solution with 0.003mM NaHCO3. |