missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Interleukin-2 / IL-2 Protein

Product Code. 15955519
Click to view available options
Quantity:
1 mg
100 μg
2 μg
20 μg
500 μg
Unit Size:
100µg
1mg
20µg
2µg
500µg
This item is not returnable. View return policy

Product Code. 15955519

Brand: enQuireBio™ QP1546100 ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_446288.1
Gene ID (Entrez) 116562
Name Interleukin-2 / IL-2 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol IL2
Biological Activity Shows bioactivity at a concentration of 2 ng/mL in the CTLL-2 Proliferation assay. The bioactivity acceptance criteria is set at 5 x 10^5 units/mg.
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag Untagged
Sequence MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAICQSI ISTMTQ
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.