missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat IL-4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£239.73 - £488.11
Specifications
Gene ID (Entrez) | 287287 |
---|---|
Molecular Weight (g/mol) | 13.5 kDa |
Name | IL-4 |
Source | E. coli-derived rat IL-4 protein Cys25-Ser147,with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18116680
|
R&D Systems™
504-RL-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 25μg |
£488.11
25µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18197820
|
R&D Systems™
504-RL-005/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 5μg |
£239.73
5µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18197220
|
R&D Systems™
504-RL-005 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 5μg |
£239.73
5µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18158270
|
R&D Systems™
504-RL-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 25μg |
£488.11
25µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The Recombinant Rat IL-4 Protein is derived from E. coli. The Recombinant Rat IL-4 Protein has been validated for the following applications: Bioactivity.Specifications
287287 | |
IL-4 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
13.5 kDa | |
E. coli-derived rat IL-4 protein Cys25-Ser147,with an N-terminal Met | |
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
Recombinant |