missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Rat IL-1ra/IL-1F3/IL1RN Protein

Product Code. p-200055636 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1mg
500 μg
Unit Size:
100µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18414902

Brand: Novus Biologicals™ NBP235225100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

IL-1ra/IL-1F3/IL1RN protein is fully biologically active when compared to standard. The ED50 as determined by inhibiting IL-1 alpha-dependent proliferation of murine D10S cells is less than 150 ng/mL, corresponding to a specific activity of >6.7 x 10^3 IU/mg in the presence of 50 pg/mL IL-1ra/IL-1F3/IL1RN
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 3557
Molecular Weight (g/mol) M.W. (Theoretical): 17.4 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/μg of IL-1ra/IL-1F3/IL1RN as determined by LAL method.
Gene Alias DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4
Gene Symbol IL1RN
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C
Research Category Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein IL-1ra/IL-1F3/IL1RN
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.