missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat IL-1 beta/IL-1F2 Protein
Description
Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells. Symons, J.A. et al. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 272. The ED50 for this effect is 0.300-1.50 ng/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in MES, NaCl, EDTA, DTT and PEG 8000. |
| Gene ID (Entrez) | 24494 |
| Molecular Weight (g/mol) | 17 kDa |
| Name | IL-1 beta/IL-1F2 |
| Quantity | 50 μg |
| Source | E. coli-derived rat IL-1 beta/IL-1F2 protein Val117-Ser268,with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?