missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IGF1 / IGF-I Protein

A DNA sequence encoding the Rattus norvegicus (Rat) IGF1 / IGF-I, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8580-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095391

  • £560.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001075946.2
IGF1 / IGF-I Protein
Research Use Only
IGF1
Recombinant Protein
E. coli
GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSA
Greater than 90% as determined by SDS-PAGE.
24482
100 μg
Not determined.
Full Length
Rat
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers