missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IGF1 / IGF-I Protein

A DNA sequence encoding the Rattus norvegicus (Rat) IGF1 / IGF-I, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8580-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16045401

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001075946.2
IGF1 / IGF-I Protein
Research Use Only
IGF1
Recombinant Protein
E. coli
GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSA
Greater than 90% as determined by SDS-PAGE.
24482
50 μg
Not determined.
Full Length
Rat
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers