missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IGF1 / IGF-I Protein
A DNA sequence encoding the Rattus norvegicus (Rat) IGF1 / IGF-I, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8580-ec-1mg
Weitere Details : Weight : 0.01000kg
Spezifikation
NP_001075946.2 | |
IGF1 / IGF-I Protein | |
Research Use Only | |
IGF1 | |
Recombinant Protein | |
E. coli | |
GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKSA | |
Greater than 90% as determined by SDS-PAGE. |
24482 | |
1 mg | |
Not determined. | |
Full Length | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |