missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat IFN-gamma Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Description
Interferon gamma Protein is Fully biologically active when compared to standard. The ED50 as determined by an anti-viral assay using murine L929 cells infected with encephalomyocarditis (EMC) virus is less than 0.1 ng/mL, corresponding to a specific activity of >1.0 x 10^7 IU/mg.
Specifications
Specifications
| Accession Number | P01581, P01581, P01581 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 1 x PBS, pH 7.4, 1 mM DTT, and 0.05 % Tween-80. |
| Gene ID (Entrez) | 3458 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 15.5 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of IFN-gamma as determined by LAL method. |
| Gene Alias | IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma |
| Gene Symbol | IFNG |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction