missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat ICOS / AILIM / CD278 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) ICOS / AILIM / CD278, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8229-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16083411

  • £2480.00 / 1mg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_072132.1
ICOS / AILIM / CD278 Protein
Research Use Only
ICOS
Recombinant Protein
E. coli
ELNDLANHRM FSFHDGGVQI SCNYPETVQQ LKMQLFKDRE VLCDLTKTKG SGNTVSIKNP MSCPYQLSNN SVSFFLDNAD SSQGSYFLCS LSIFDPPPFQ EKNLSGGYLL IYESQLCCQL KLWLP
Greater than 90% as determined by SDS-PAGE.
64545
1 mg
Not determined.
Extracellular Domain
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers