missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Rat HO-1/HMOX1/HSP32 His Protein

Product Code. p-7249117 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
200 μg
50 μg
Unit Size:
100µg
200µg
50µg
This item is not returnable. View return policy

Product Code. 18345734

Brand: Novus Biologicals™ NBP31832850UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE, Western Blot
Formulation 50mM Tris/HCl pH7.5, 5mM Bme, 0.15NaCl, 10% glycerol
Gene ID (Entrez) 3162
Molecular Weight (g/mol) M.W. Theoretical: 32 kDa
Quantity 50 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32
Gene Symbol HMOX1
Research Category Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism
Purity or Quality Grade >85%
Protein HO-1/HMOX1/HSP32
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.